Learn More
Rat IgG1 Kappa Light Chain, Clone: RG1, Isotype control, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1433220.05MG
 View more versions of this product
Catalog No. NBP1433225M
Add to cart
Description
Description
IgG1 Kappa Light Chain Monoclonal antibody specifically detects IgG1 Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Control, Neutralization.
Specifications
Specifications
IgG1 Kappa Light Chain
RG1
Unconjugated
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Rat
0.05 mg
Primary
The monoclonal rat IgG1, kappa immunoglobulin is useful as an isotype control for rat IgG1 antibodies.
ELISA, Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000
Liquid
IgG1 Kappa
Store at 4C in the dark.
Monoclonal
Rat