Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2a Kappa Light Chain, Clone: R2a, APC, Isotype control, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1430050.05MG

Catalog No. NBP14300505


Add to cart

Description

Description

IgG2a Kappa Light Chain Monoclonal antibody specifically detects IgG2a Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

IgG2a Kappa Light Chain
R2a
Flow Cytometry 1:10 - 1:1000
Conjugated
IgG2a Kappa
Store at 4C in the dark.
Monoclonal
Rat
Flow Cytometry
APC
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Rat
0.05 mg
Primary
The monoclonal rat IgG2a, kappa immunoglobulin is useful as an isotype control for rat IgG2a antibodies.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit