Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IgG2a Kappa Light Chain, Clone: R2a, FITC, Isotype control, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP1439590.05MG
Description
IgG2a Kappa Light Chain Monoclonal specifically detects IgG2a Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.Specifications
IgG2a Kappa Light Chain | |
Monoclonal | |
FITC | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
0.05 mg | |
The monoclonal rat IgG2a, kappa immunoglobulin is useful as an isotype control for rat IgG2a antibodies. | |
Store at 4C in the dark. |
Flow Cytometry | |
R2a | |
Flow Cytometry 1:10 - 1:1000 | |
Rat | |
Primary | |
Rat | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction