Learn More
Rat IgG2a Kappa Light Chain, Clone: R2a, PE-Cy7, Isotype control, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1429450.1MG
 View more versions of this product
Catalog No. NBP14294501
Add to cart
Description
Description
IgG2a Kappa Light Chain Monoclonal antibody specifically detects IgG2a Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
IgG2a Kappa Light Chain
R2a
PE-Cyanine7
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Rat
IgG purified
Store at 4C in the dark.
Monoclonal
Rat
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10 - 1:1000
IgG Purified
IgG2a Kappa
0.1 mg
Primary
The monoclonal rat IgG2a, kappa immunoglobulin is useful as an isotype control for rat IgG2a antibodies.