Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2a kappa, Super Bright 600, clone: eBR2a, Isotype Control, eBioscience™

Rat Isotype Control

$161.00 - $334.00

Specifications

Antigen Rat IgG2a kappa
Clone eBR2a
Target Species Not Applicable
Conjugate Super Bright 600
Isotype IgG2a, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
63-432-180
Encompass
View Documents
Invitrogen
63432180
25μg
Each for $161.00
Add to cart
 
63-432-182
Encompass
View Documents
Invitrogen
63432182
100μg
Each for $334.00
Add to cart
 
Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin.Applications Reported: This eBR2a antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBR2a antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use the isotype control at the same concentration as the experimental antibody.Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100μL of cell sample + 100μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 2-8°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorochrome performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser
Specifications

Specifications

Rat IgG2a kappa
Not Applicable
IgG2a, kappa
Liquid
Isotype Control
4°C, store in dark, DO NOT FREEZE!
eBR2a
Super Bright 600
Control, Flow Cytometry
Control
PBS with BSA and 0.09% sodium azide; pH 7.2
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit