Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IgG2b Kappa Light Chain, Clone: 149/10H5, Biotin, Isotype control, Novus Biologicals™
Rat Monoclonal Antibody
$191.70 - $554.50
Specifications
Antigen | IgG2b Kappa Light Chain |
---|---|
Clone | 149/10H5 |
Dilution | Flow Cytometry 1:10 - 1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14356305
|
Novus Biologicals
NBP1435630.05MG |
0.05 mg |
Each for $191.70
|
|
|||||
NBP14356301
|
Novus Biologicals
NBP1435630.1MG |
0.1 mg |
Each for $301.00
|
|
|||||
NBP1435635
|
Novus Biologicals
NBP1435630.5MG |
0.5 mg |
Each for $554.50
|
|
|||||
Description
IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.Specifications
IgG2b Kappa Light Chain | |
Flow Cytometry 1:10 - 1:1000 | |
Monoclonal | |
Rat | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
Primary | |
The monoclonal rat IgG2b, kappa immunoglobulin is useful as an isotype control for rat IgG2b antibodies. |
149/10H5 | |
Flow Cytometry | |
Biotin | |
Rat | |
IgG2b κ | |
Store at 4C in the dark. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title