Learn More
Rat IgG2b Kappa Light Chain, Clone: 149/10H5, Isotype control, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1433230.5MG
 View more versions of this product
Catalog No. NBP1433235
Add to cart
Description
Description
IgG2b Kappa Light Chain Monoclonal antibody specifically detects IgG2b Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In vitro assay, Control.
Specifications
Specifications
IgG2b Kappa Light Chain
149/10H5
Unconjugated
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Rat
0.5 mg
Primary
The monoclonal rat IgG2b, kappa immunoglobulin is useful as an isotype control for rat IgG2b antibodies.
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10 - 1:1000
Liquid
IgG2b Kappa
Store at 4C in the dark.
Monoclonal
Rat