Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2b kappa, Super Bright 645, clone: eB149/10H5, Isotype Control, eBioscience™ Non-distribution product as customer accommodation.

Rat Isotype Control

$150.00 - $312.00

Specifications

Antigen Rat IgG2b kappa
Clone eb149/10H5
Target Species Not Applicable
Conjugate Super Bright 645
Isotype IgG2b, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
64403180 Non-distribution product as customer accommodation. View Documents Invitrogen
64403180
25μg Each for $150.00
Add to cart
 
64403182 Non-distribution product as customer accommodation. View Documents Invitrogen
64403182
100μg Each for $312.00
Add to cart
 
Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2b, kappa, isotype control has been reported for use as a control for immunofluorescent staining, flow cytometric analysis, immunohistochemistry and immunocytochemistry. Applications Tested: This rat IgG2b, kappa, isotype control has been tested by immunofluorescent staining and flow cytometric analysis of mouse splenocyte suspensions and can be used at the same concentration as the experimental antibody. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100µL of cell sample + 100µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser
Specifications

Specifications

Rat IgG2b kappa
Not Applicable
IgG2b, kappa
Liquid
Isotype Control
4°C, store in dark, DO NOT FREEZE!
eb149/10H5
Super Bright 645
Control, Flow Cytometry
Control
PBS with BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit