Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2b, κ Isotype Control, BUV615, BD Biosciences™
Monoclonal Isotype Control
Supplier: BD Biosciences 613007
Description
IgG2b, κ Monoclonal antibody specifically detects IgG2b, κ. It is validated for Flow Cytometry, ControlSpecifications
IgG2b, κ | |
Monoclonal | |
Brilliant Ultraviolet 615 | |
Rat IgG2b kappa Isotype Control | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Flow Cytometry | |
R35-38 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
50 μg | |
Isotype Control (Primary) | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction