Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IgM Kappa Light Chain, Clone: RGM, Isotype control, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP1433200.5MG
Description
IgM Kappa Light Chain Monoclonal specifically detects IgM Kappa Light Chain in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Control.Specifications
IgM Kappa Light Chain | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
Rat | |
Primary | |
Rat | |
Liquid |
Flow Cytometry | |
RGM | |
Unconjugated | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
0.5 mg | |
The monoclonal rat IgM, kappa immunoglobulin is useful as an isotype control for rat IgM antibodies. | |
Store at 4C in the dark. | |
IgM κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction