Learn More
Novus Biologicals™ Rat IL-1 beta/IL-1F2 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192702
Catalog No. NBP192702
Add to cart
Description
Description
The Rat IL-1 beta/IL-1F2 ELISA Kit (Colorimetric) measures rat IL-1 beta/IL-1F2 in cell culture supernatant, serum.
Specifications
Specifications
31.3 - 2,000pg/mL
3553
ELISA Kit
4.0pg/mL
Rat (Rt)
IL-1 beta/IL-1F2
IL1B
ELISA
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to rat IL-1beta, Biotin-Conjugate anti-rat IL-1beta polyclonal antibody, Streptavidin-HRP, Rat IL-1beta Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Apoptosis, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta