Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit
Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$3804.80
Specifications
Assay Range | 31.20 to 2000 pg/mL |
---|---|
Assay Sensitivity | 5 pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
Product Type | Solid Phase Sandwich ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
SRLB00
|
R&D Systems™
SRLB00 |
6 Plates |
Each for $3,804.80
|
|
|||||
RLB00
|
R&D Systems™
RLB00 |
2 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
The Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat IL-1 beta/IL-1F2.Specifications
31.20 to 2000 pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Rat IL-1 beta | |
ELISA | |
Cell Culture Supernates (50 μL), Serum (17 μL), EDTA Plasma (17 μL) | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1 b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
5 pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA), Serum | |
Rat | |
24494 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title