Learn More
Raybiotech IncSupplier Diversity Partner Rat IL-10 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELRIL101
For cell culture supernatants, plasma, and serum samples. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with any of the cytokines tested: rat CINC-2, CINC-3, CNTF, IL-1 alpha, IL-1 beta, IL-4, IL-6, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha, VEGF.
Catalog No. 50-195-0255