Learn More
Novus Biologicals™ Rat IL-13 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Supplier: Novus Biologicals™ NBP306796
This item is not returnable.
View return policy
Specifications
15.63 to 1000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat IL-13 in samples. No significant cross-reactivity or interference between Rat IL-13 and analogues was observed. | |
3596 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Rat |
9.38 pg/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
IL13 | |
Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600 |