Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat IL-17/IL-17A ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP192705
Description
The Rat IL-17/IL-17A ELISA Kit (Colorimetric) measures rat IL-17/IL-17A in cell culture supernatant, serum, plasma (edta, citrate, heparin).Specifications
1.6 - 100pg/mL | |
ELISA Kit | |
Rat | |
3605 | |
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-17A, Biotin-Conjugate anti-rat IL-17A monoclonal antibody, Streptavidin-HRP, Rat IL-17A Standard lyophilized, 200 pg/ml upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films | |
Apoptosis, Cytokine Research, Immunology, Innate Immunity | |
CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8 |
1.0pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
ELISA | |
IL17A | |
96 Tests | |
Storage of components varies. See protocol for specific instructions. | |
IL-17/IL-17A |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction