Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Rat IL-2 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192704
Catalog No. NBP192704
Description
Description
The Rat IL-2 ELISA Kit (Colorimetric) measures rat IL-2 in cell culture supernatant, serum.
Specifications
Specifications
62.5 - 2,000pg/mL
3558
ELISA Kit
11.0pg/mL
Rat (Rt)
IL-2
IL2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-2, Biotin-Conjugate anti-rat IL-2 monoclonal antibody, Streptavidin-HRP, Rat IL-2 Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity
aldesleukin, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF