Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IL-3 (aa 27-169), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$244.00 - $3850.00

Specifications

Recombinant Yes
Name Rat IL-3 (aa 27-169)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87928 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87928
10 µg
Each for $244.00
Add to cart
 
PIRP87929 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87929
25 µg
Each for $481.50
Add to cart
 
PIRP87930 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87930
100 µg
Each for $1,278.00
Add to cart
 
PIRP87931 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87931
500 µg
Each for $3,850.00
Add to cart
 
Description

Description

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative; pH 7.2
E. coli
-20°C
P04823
IL3
>95%
Rat IL-3 (aa 27-169)
200 µg/mL
Liquid
BP/BPA; BPA; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; Hematopoietic growth factor; Il3; IL-3; ILN; Interleukin; interleukin 3; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; MULTI-CSF; multilineage-colony-stimulating factor; multipotential colony-stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF
RUO
Rat
24495
16.3 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit