Learn More
Abnova™ Rat Il13 (P42203) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6087
Catalog No. 89-948-646
Add to cart
Description
Description
Sequence: TPGPVRRSTSPPVALRELIEELSNITQDQKTSLCNSSIVWSVDLTAGGFCAALESLTNISSCNAIHRTQRILNGLCNQKASDVASSPPDTKIEVAQFISKLLNYSKQLFRYGH
Specifications
Specifications
P42203
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
12.3kDa
E. coli expression system
10 ug
Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
Il13
Rat ;Rat
Yes
E. coli expression system
Lyophilized
116553
Il13 (Rat) Recombinant Protein
0.98
TPGPVRRSTSPPVALRELIEELSNITQDQKTSLCNSSIVWSVDLTAGGFCAALESLTNISSCNAIHRTQRILNGLCNQKASDVASSPPDTKIEVAQFISKLLNYSKQLFRYGH
RUO
Il13
The ED50 determined by the dose-dependent stimulation of the proliferation of TF-1 cells is < 5.0ng/ml, corresponding to a specific activity of > 2 x 105 units/mg.
Escherichia coli
None