Learn More
Abnova™ Rat Il17f Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4846
Catalog No. 89-948-697
Add to cart
Description
Description
Sequence: MARRNPKVGLSALQKAGNCPPLEDNSVRVDIRIFNQNQGISVPRDFQNRSSSPWDYNITRDPDRFPSEIAEAQCRHSGCINAQGQEDGSMNSVPIQQEILVLRREPQGCSNSFRLEKMLIKVGCTCVTPIVHHAA
Specifications
Specifications
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
30kDa
Escherichia coli expression system
25 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Il17f
Recombinant rat Il17f is fully bioactive when compared to a known standards. The ED50, as determined by the dose-dependant induction of IL-6 in NIH3T3 cells, is 9.18ng/mL.
Yes
Q5BJ95
Lyophilized
301291
Il17f (Rat) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel stained with Coomassie Blue
MARRNPKVGLSALQKAGNCPPLEDNSVRVDIRIFNQNQGISVPRDFQNRSSSPWDYNITRDPDRFPSEIAEAQCRHSGCINAQGQEDGSMNSVPIQQEILVLRREPQGCSNSFRLEKMLIKVGCTCVTPIVHHAA
RUO
MGC114402
Il17f
Escherichia coli
None