Learn More
Abnova™ Rat Il2 (P17108, 134 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5139
Catalog No. 89-948-796
Add to cart
Specifications
Specifications
P17108
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
15.3kDa
Escherichia coli expression system
20 ug
RUO
Il2
The ED50 as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is 0.4ng/ml, corresponding to a specific activity of ≥ 2.5 x 106 units/mg.
Yes
Escherichia coli expression system
Lyophilized
116562
Il2 (Rat) Recombinant Protein
≥98% by SDS-PAGE and HPLC
Store at -20°C on dry atmosphere. After reconstitution with sterilized 20 mM acetic acid, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Il2
Escherichia coli
None