Learn More
Abnova™ Rat Il25 (XP_577423, 145 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5148
Catalog No. 89-948-844
Add to cart
Specifications
Specifications
XP_577423
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
35.5kDa
Escherichia coli expression system
10 ug
RUO
RGD1561632
Il25
Escherichia coli
None
Escherichia coli expression system
Lyophilized
501996
Il25 (Rat) Recombinant Protein
≥95% by SDS-PAGE
Store at -20°C on dry atmosphere. After reconstitution with sterilized 10 mM HCl, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Il25
Recombinant rat Il25 has full activity when compared to standard. Activity was measured by the dose-dependent induction of mouse Il5 production in cultured mouse splenocytes which is typically 20-100ng/mL.
Yes