Learn More
Abnova™ Rat Il3 (P97688) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6100
Catalog No. 89-948-879
Add to cart
Description
Description
Sequence: MISDRGSDAHHLLRTLDCRTIALEILVKLPYPQVSGLNNSDDKANLRNSTLRRVNLDEFLKSQEEFDSQDTTDIKSKLQKLKCCIPAAASDSVLPGVYNKDLDDFKKKLRFYVIHLKDLQPVSVSRPPQPTSSSDNFRPMTVEC
Specifications
Specifications
P97688
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
16.3kDa
E. coli expression system
20 ug
Store at -20°C. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. For extended storage, it is recommended to be further diluted in buffer containing carrier protein (e.g. 0.1% BSA) and stored in aliquots at -20°C to -80°C.
<0.1ng/μg (<1 EU/μg)
Il3
Rat ;Rat
Yes
E. coli expression system
Lyophilized
24495
Il3 (Rat) Recombinant Protein
0.98
MISDRGSDAHHLLRTLDCRTIALEILVKLPYPQVSGLNNSDDKANLRNSTLRRVNLDEFLKSQEEFDSQDTTDIKSKLQKLKCCIPAAASDSVLPGVYNKDLDDFKKKLRFYVIHLKDLQPVSVSRPPQPTSSSDNFRPMTVEC
RUO
Il3
The ED50 determined by the stimulation of the proliferation with murine MC-9 cells is < 10ng/ml, corresponding to a specific activity of > 1.0 x 105 units/mg.
Escherichia coli
None