Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Rat Il7 (P56478, 130 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P5149
Specifications
P56478 | |
Lyophilized | |
15.0kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
Il7 | |
E. coli | |
None | |
Lyophilized |
Functional Study, SDS-PAGE | |
25647 | |
Il7 (Rat) Recombinant Protein | |
10 ug | |
RUO | |
Il7 | |
The ED50 as determined by the dose-dependent stimulation of the proliferation of murine 2e8 cells is < 2ng/mL, corresponding to a specific activity of > 5 x 106 units/mg. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and HPLC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction