Learn More
Raybiotech IncSupplier Diversity Partner Rat Jagged 1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELRJAGGED11
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 73 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat Jagged 1.
Catalog No. 50-195-0274