Learn More
Novus Biologicals™ Rat Bradykinin ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP269971
Catalog No. NBP269971
Add to Cart
Description
Description
The Rat Kininogen/Kininostatin/Bradykinin ELISA Kit (Colorimetric) measures rat Kininogen/Kininostatin/Bradykinin in serum, plasma and other biological fluids.
Specifications
Specifications
1.25 - 80 ng/mL
This kit recognizes Rat BK in samples. No significant cross-reactivity or interference between Rat BK and analogues was observed.
Biological Fluids, Plasma, Serum
ELISA
KNG1
1 Kit
Kininogen/Kininostatin/Bradykinin
0.75 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat BK in samples. No significant cross-reactivity or interference between Rat Bradykinin and analogues was observed.
3827
Rat
Storage of components varies. See protocol for specific instructions.
Product Suggestions
Product Suggestions