Learn More
Novus Biologicals™ Rat KISS1 ELISA Kit (Colorimetric)
Rat KISS1 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: Novus Biologicals™ NBP276460
Description
The Rat KISS1 ELISA Kit (Colorimetric) measures rat KISS1 in serum, plasma and other biological fluids.Specifications
78.13 to 5000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat KISS1 in samples. No significant cross-reactivity or interference between Rat KISS1 and analogues was observed. | |
KISS1 | |
1 Kit |
46.88 pg/mL | |
Biological Fluids, Plasma, Serum | |
3814 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |