Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Rat Kitlg Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P4851
Description
Sequence: MQEICRNPVTDNVKDITKLVANLPNDYMITLNYVAGMDVLPSHCWLRDMVTHLSVSLTTLLDKFSNISEGLSNYSIIDKLGKIVDDLVACMEENAPKNVKESLKKPETRNFTPEEFFSIFNRSIDAFKDFMVASDTSDCVLSSTLGPEKDSRVSVTKPFMLPPVASpecifications
P21581 | |
Lyophilized | |
18.4kDa | |
Escherichia coli expression system | |
10 ug | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized 10 mM acetic acid, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1 EU/μg | |
Kitlg | |
The activity is determined by the ability to induce proliferation of TF-1 cells and is 10-15ng/mL. | |
Recombinant | |
Escherichia coli expression system |
Functional Study, SDS-PAGE | |
60427 | |
Kitlg (Rat) Recombinant Protein | |
1 ug/lane in 4-20% Tris-Glycine gel stained with Coomassie Blue | |
MQEICRNPVTDNVKDITKLVANLPNDYMITLNYVAGMDVLPSHCWLRDMVTHLSVSLTTLLDKFSNISEGLSNYSIIDKLGKIVDDLVACMEENAPKNVKESLKKPETRNFTPEEFFSIFNRSIDAFKDFMVASDTSDCVLSSTLGPEKDSRVSVTKPFMLPPVA | |
RUO | |
Kitl/Mgf/SCF | |
Kitlg | |
E. coli | |
None | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction