Learn More
Raybiotech IncSupplier Diversity Partner L-Series Rat Antibody Array 90 Membrane Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner AARBLM1A2
Detects 90 Rat Proteins. Suitable for Cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAR-BLM-1A-2
Catalog No. 50-194-8771