Learn More
Raybiotech IncSupplier Diversity Partner Rat MAG ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELRMAG1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 10 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat MAG.
Catalog No. 50-195-0281