Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat MBL-2 ELISA Kit Non-distribution product as customer accommodation.

Rat MBL-2 quantitates rat MBL-2 in serum, plasma, supernatant.

$491.00 - $2205.00

Specifications

Regulatory Status Research Use Only
Storage Requirements 2°C to 8°C
Target MBL2
Product Type ELISA
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERA38RBX5 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA38RBX5
5 x 96 Tests
Each for $2,205.00
Add to cart
 
ERA38RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA38RB
96 Tests
Each for $491.00
Add to cart
 
Description

Description

Rat MBL-2 quantitates rat MBL-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat MBL-2.

MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
Specifications

Specifications

Research Use Only
MBL2
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
4 hr 45 min
Serum, Plasma, Supernatant
2°C to 8°C
ELISA
26 pg/mL
1 hr 20 min
Supernatant, 50 µL; Plasma, 50 µL; Serum, 50 µL
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit