Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Rat Met (X96786, 958 a.a. - end) Partial Recombinant Protein expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova™ P5150
Specifications
X96786 | |
Liquid | |
24553 | |
Met (Rat) Recombinant Protein | |
≥90% by SDS-PAGE | |
5 ug | |
RUO | |
Met | |
The specific activity of Met was determined to be 46nmol/min/mg. | |
Yes | |
Insect cell (Sf9) expression system |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
74.0kDa | |
Insect cell (Sf9) expression system | |
SDS-PAGE Result | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Hgfr | |
Met | |
Insect | |
None |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title