Learn More
Novus Biologicals™ Rat MUC2 ELISA Kit (Colorimetric)
Rat MUC2 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276701
Catalog No. NBP276701
Add to cart
Description
Description
The Rat MUC2 ELISA Kit (Colorimetric) measures rat MUC2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
MUC2
Biological Fluids, Plasma, Serum
4583
0.188 ng/mL
Rat
1 Kit