Learn More
Invitrogen™ Rat Neuropilin-2 ELISA Kit
ELISA
$700.00 - $5890.00
Specifications
Accession Number | 1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2 A(17); neuropilin-2 A(22); neuropilin-2 b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2 |
---|---|
Assay Range | 0.61 to 150 ng/mL |
Assay Sensitivity | 0.61 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ERA43RBX5
![]() |
Invitrogen™
ERA43RBX5 |
5 x 96 Tests |
Each for $3,285.00
|
|
|||||
ERA43RB
![]() |
Invitrogen™
ERA43RB |
96 Tests |
Each for $700.00
|
|
|||||
50-254-5428
![]() |
Invitrogen™
ERA43RBX10 |
10 x 96 Tests |
Each for $5,890.00
|
|
|||||
Description
Rat Neuropilin-2 quantitates rat Neuropilin-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Neuropilin-2.
This gene encodes a member of the neuropilin family of receptor proteins. The encoded transmembrane protein binds to SEMA3C protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C) and SEMA3F protein (sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3F), and interacts with vascular endothelial growth factor (VEGF). This protein may play a role in cardiovascular development, axon guidance, and tumorigenesis. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Specifications
1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2 A(17); neuropilin-2 A(22); neuropilin-2 b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2 | |
0.61 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
NRP2 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
Neuropilin 2 | |
4 hrs 45 mins |
0.61 to 150 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
81527 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.