Learn More
Crystal Chem IncSupplier Diversity Partner Rat NGAL ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80687
The Rat NGAL ELISA is used to quantify levels of rat NGAL in serum, plasma, and urine using only a 2 µL sample volume. Highlights: Kit uses sample size of only 2 µL. Highly precise with low CV's. Dynamic range of 0.25 - 8 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7814