Learn More
Novus Biologicals Rat NK1R ELISA Kit (Colorimetric)
This kit recognizes Rat TACR1 in samples.
Manufacturer: Novus Biologicals NBP280293
Description
No significant cross-reactivity or interference between Rat TACR1 and analogues was observed.Specifications
P14600, P14600 | |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
1 Kit | |
Serum, plasma, and other biological fluids |
125 to 8000 pg/mL | |
6869 | |
75 pg/mL | |
Rat | |
TACR1 | |
NK1R |