Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Notch-2 ELISA Kit Non-distribution product as customer accommodation.

Rat Notch-2 quantitates rat Notch-2 in serum, plasma, supernatant.

$491.00 - $2205.00

Specifications

Regulatory Status Research Use Only
Storage Requirements 2°C to 8°C
Target NOTCH2
Product Type ELISA
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERA45RBX5 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA45RBX5
5 x 96 Tests
Each for $2,205.00
Add to cart
 
ERA45RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA45RB
96 Tests
Each for $491.00
Add to cart
 
Description

Description

Rat Notch-2 quantitates rat Notch-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Notch-2.

Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.
Specifications

Specifications

Research Use Only
NOTCH2
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
4 hr 45 min
Serum, Plasma, Supernatant
2°C to 8°C
ELISA
0.14 ng/mL
1 hr 20 min
Supernatant, 50 µL; Plasma, 50 µL; Serum, 50 µL
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit