Learn More
Novus Biologicals™ Rat NT-4 ELISA Kit (Colorimetric)
Rat NT-4 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: Novus Biologicals™ NBP276770
Description
The Rat NT-4 ELISA Kit (Colorimetric) measures rat NT-4 in serum, plasma and other biological fluids.Specifications
31.25 to 2000 pg/mL | |
ELISA Kit (Colorimetric) | |
4909 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
18.75 pg/mL | |
Biological Fluids, Plasma, Serum | |
NTF4 | |
1 Kit |