Learn More
Zyagen LabsSupplier Diversity Partner Rat olfactorytory epithelium frozen sections are made of normal healthy tissue of Sprague Dawley rat single donor. Sections of 10 microns are mounted on positively charged slides. Ideal for immunohistochemistry & in situ hybridization. 10 slides/PK. DFS Item
Manufacturer:  Zyagen LabsSupplier Diversity Partner RF606
Zyagen Frozen tissue sections are made of normal healthy tissues which are freshly harvested of single donor and snapped frozen in OCT blocks without any fixative. Blocks are sectioned by cryostat at a thickness of 7 or 10microns and sections mounted on positively charged slides (2 sections per slide). Frozen tissue sections are prepared in RNase- and protease-free environment and are ideal for immunohistochemistry and in situ hybridization. Tissue sections are sold in a unit of 10 slides. Frozen sections are shipped in vacuum sealed slide boxes or slide mailers on dry ice and are good for one year if properly stored at-80oC. Quality of tissue morphology is tested using H&E staining.
Catalog No. 50-172-0002