Learn More
Novus Biologicals Rat Orexin B/Hypocretin-2 ELISA Kit (Colorimetric)
This kit recognizes Rat OXB in samples.
Manufacturer: Novus Biologicals NBP280236
Description
No significant cross-reactivity or interference between Rat OXB and analogues was observed.Specifications
0.156 to 10 ng/mL | |
3060 | |
0.094 ng/mL | |
Rat | |
HCRT | |
Orexin B/Hypocretin-2 |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
1 Kit | |
Serum, plasma, and other biological fluids |