Learn More
BioworldSupplier Diversity Partner RAT PARVALBUMIN (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220606731
Recombinant Rat Parvalbumin (PV) Source: E.Coli Description: Parvalbumin (PV or PVALB or oncomodulin) is a high affinity low mol wt (~12 kDa) cytoplasmic protein belonging to the troponin C superfamily. It is expressed in high levels...
Catalog No. 50-253-769