Learn More
Novus Biologicals™ Rat proANP ELISA Kit (Colorimetric)
Rat proANP ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276773
Catalog No. NBP276773
Add to cart
Description
Description
The Rat proANP ELISA Kit (Colorimetric) measures rat proANP in serum, plasma and other biological fluids.
Specifications
Specifications
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
NPPA
Biological Fluids, Plasma, Serum
4878
0.188 ng/mL
Rat
1 Kit