Learn More
Invitrogen™ Rat Prolactin ELISA Kit
ELISA
$682.65 - $5677.65
Specifications
Accession Number | AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL |
---|---|
Assay Range | 0.41 to 100 ng/mL |
Assay Sensitivity | 0.41 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ERA50RBX5
![]() |
Invitrogen™
ERA50RBX5 |
5 x 96 Tests |
Each
for $3,171.65
Save 4% Reg : |
|
|||||
ERA50RB
![]() |
Invitrogen™
ERA50RB |
96 Tests |
Each
for $682.65
Save 4% Reg : |
|
|||||
50-254-5420
![]() |
Invitrogen™
ERA50RBX10 |
10 x 96 Tests |
Each
for $5,677.65
Save 4% Reg : |
|
|||||
Description
Rat Prolactin quantitates rat Prolactin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Prolactin.
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.Specifications
AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
0.41 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
PRL | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
Prolactin | |
4 hrs 45 mins |
0.41 to 100 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
24683 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.