Learn More
Novus Biologicals™ Rat SDF2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275379
Catalog No. NBP275379
Add to cart
Description
Description
The Rat SDF2 ELISA Kit (Colorimetric) measures rat SDF2 in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat SDF-2 in samples. No significant cross-reactivity or interference between Rat SDF-2 and analogues was observed.
6388
9.38 pg/mL
SDF2
SDF2
Biological Fluids, Plasma, Serum