Learn More
Novus Biologicals™ Rat Serpin G1/C1 Inhibitor ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP270030
Catalog No. NBP270030
Add to Cart
Description
Description
The Rat Serpin G1/C1 Inhibitor ELISA Kit (Colorimetric) measures rat Serpin G1/C1 Inhibitor in serum, plasma and other biological fluids.
Specifications
Specifications
7.81 - 500 ng/mL
This kit recognizes Rat C1INH in samples. No significant cross-reactivity or interference between Rat C1INH and analogues was observed.
Biological Fluids, Plasma, Serum
710
Rat
Storage of components varies. See protocol for specific instructions.
4.69 ng/mL
ELISA Kit (Colorimetric)
ELISA
SERPING1
1 Kit
Serpin G1/C1 Inhibitor
Recommendations
Recommendations