Learn More
Novus Biologicals™ Rat SLC6A3/DAT1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP267273
Description
The Rat SLC6A3/DAT1 ELISA Kit (Colorimetric) measures rat SLC6A3/DAT1 in serum, plasma and other biological fluids.Specifications
0.31 - 20 ng/mL | |
This kit recognizes Rat DAT in samples. No significant cross-reactivity or interference between Rat DAT and analogues was observed. | |
6531 | |
Rat | |
1 Kit | |
Storage is content dependent. |
0.19 ng/mL | |
ELISA | |
SLC6A3 | |
ELISA Kit (Colorimetric) | |
Biological Fluids,Plasma,Serum | |
SLC6A3/DAT1 |