Learn More
Novus Biologicals™ Rat Smad7 ELISA Kit (Colorimetric)
Rat Smad7 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276628
Catalog No. NBP276628
Add to cart
Description
Description
The Rat Smad7 ELISA Kit (Colorimetric) measures rat Smad7 in serum, plasma and other biological fluids.
Specifications
Specifications
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat Smad7/MADH7 in samples. No significant cross-reactivity or interference between Rat Smad7/MADH7 and analogues was observed.
Rat
1 Kit
4092
0.188 ng/mL
Storage is content dependent.
SMAD7
Biological Fluids, Plasma, Serum