Learn More
Novus Biologicals™ Rat Surfactant Protein A ELISA Kit (Colorimetric)
Rat Surfactant Protein A ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276694
Catalog No. NBP276694
Add to cart
Description
Description
The Rat Surfactant Protein A ELISA Kit (Colorimetric) measures rat Surfactant Protein A in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 to 2000 pg/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
SFTPA1
Biological Fluids, Plasma, Serum
653509
18.75 pg/mL
Rat
1 Kit