Learn More
Novus Biologicals™ Rat Tau [p Ser610] ELISA Kit (Colorimetric)
Rat Tau [p Ser610] ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276716
Catalog No. NBP276716
Add to cart
Description
Description
The Rat Tau [p Ser610] ELISA Kit (Colorimetric) measures rat Tau in serum, plasma and other biological fluids.
Specifications
Specifications
15.625 to 1000 pg/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
MAPT
Biological Fluids, Plasma, Serum
4137
9.375 pg/mL
Rat
1 Kit