Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat Thyroxine ELISA Kit (Colorimetric)
Assay has high sensitivity and specificity for detection of rat T4
Supplier: Novus Biologicals™ NBP2601641KIT
Description
The Rat Thyroxine ELISA Kit (Colorimetric) measures rat Thyroxine in serum, plasma, cell culture supernates, tissue homogenates, cell lysates.Specifications
20ng/mL to 320ng/mL | |
ELISA Kit (Colorimetric) | |
For detection of rat T4 | |
ELISA | |
1 Kit | |
50 to 100 μL | |
T4 |
20ng/mL | |
Cell Culture Supernatant, Cell Lysates, Plasma, Serum, Tissue Homogenates | |
Rat | |
Instruction manual, Stop Solution, Standard, Wash Buffer (20 x concentrate), Substrate A, Substrate B, Conjugate, HRP-avidin, Adhesive Strip (For 96 wells), Assay plate | |
Signal Transduction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Thyroxine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction