Learn More
Novus Biologicals™ Rat TLR9 ELISA Kit (Colorimetric)
Rat TLR9 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276577
Catalog No. NBP276577
Add to cart
Description
Description
The Rat TLR9 ELISA Kit (Colorimetric) measures rat TLR9 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 to 10 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat TLR9 in samples. No significant cross-reactivity or interference between Rat TLR9 and analogues was observed.
Rat
1 Kit
54106
0.10 ng/mL
Storage is content dependent.
TLR9
Biological Fluids, Plasma, Serum